Learn More
Apexbio Technology LLC hemagglutinin precursor (114-122) amide [Influenza A virus] 25mg
SDP
Supplier:  Apexbio Technology LLC A114225
Encompass_Preferred
Hemagglutinin precursor 114-122 amide is a synthetic peptide fragment derived from the hemagglutinin glycoprotein of Influenza A virus It is designed to present antigenic determinants that facilitate the analysis of peptide-MHC interactions and characterization of influenza-specific immune responses Hemagglutinin precursor 114-122 amide exerts its biological activity primarily by eliciting immune responses against Influenza A virus supporting studies on cytotoxic T-cell reactivity and antigenicity Based on these properties hemagglutinin precursor 114-122 amide holds research potential in immunological assays vaccine antigenicity studies and investigations of antiviral immune responses
Catalog No. 50-000-95536