Learn More
Microscope World Fein Optic RB40 Hematology Plan Achromat Microscope
SDP
Supplier:  Microscope World RB40HEMA2
Encompass_Preferred
Quintuple objective turret holds the included DIN Infinity Corrected Optical System (ICOS) ObjectivesPlan Achromat 10x, NA 0.25, WD 12.1mmPlan Achromat 40x, NA 0.65, WD 0.36mmPlan Achromat 50x, NA 0.95, WD 0.19mm, Oil ImmersionPlan Achromat 100x, NA 1.25, WD 0.18mm, Oil ImmersionTrinocular head. Beam splitter with positions 100-0 and 0-100. FPL-WF 10x Widefield, high eyepoint eyepieces, FN22.Magnification 100x, 400x, 500x, and 1000x.Double-layer mechanical stage, 175mm x 145mm with right-hand ergonomic controls. Specimen holder allows for either one or two slides. X-Y Axis travel of 76mm x 50mm.3W LED transmitted Koehler light system gives a bright and uniform field for each magnification. Telecentric Koehler illumination system provides NA 1.2/0.22 swing-out condenser for use with 2x-100x objectives. Coaxial coarse and fine focus adjustment with graduations of 2um. 9.8in (250mm) W x 18.2in (463mm) D x 16in (408mm) HWarranty- 5 Years.
Catalog No. NC2965679
Only null left
Add to Cart
Request a Quote