Learn More
Medchemexpress LLC HY-B0754 100mg Medchemexpress, Hematoporphyrin CAS:14459-29-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0754100MG
Encompass_Preferred
Medchemexpress, HY-B0754 100mg Hematoporphyrin CAS:14459-29-1 Hematoporphyrin (Hematoporphyrin IX), a photosensitizer, is a substrate for affinity chromatography of heme-binding proteins. Hematoporphyrin can induce apoptosis in U87 glioma cells and decrease tumor growth in vivo when exposed to red light. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2657