Learn More
TARGETMOL CHEMICALS INC Hematoporphyrin monomethyl eth
SDP
Supplier:  TARGETMOL CHEMICALS INC T389705MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Hematoporphyrin monomethyl ether is a porphyrin photosensitizer that can be used in studies of port wine stains.
Catalog No. 50-287-9163