Learn More
Biogenex Laboratories HEMATOXYLIN 250ML
SDP
Supplier:  Biogenex Laboratories HK1009K
Encompass_Preferred
Hematoxylin,250ml, Mayer's (non-alcohol-based),Biological Stain, suitable for diagnostic histopathology, laboratory, and research use
Catalog No. NC9868235