Learn More
Leica Biosystems HEMATOXYLIN 560MX 500 ML 4/CS
Supplier:  Leica Biosystems 3801575
Encompass_Preferred
SelecTech is a hematoxylin and eosin staining system consisting of Hematoxylin 560, Blue Buffer 8, Define, Alcoholic Eosin Y 515 and Eosin Trichrome 515.SelecTech standardizes the reagents and stains used in the hematoxylin and eosin staining procedure. The built-in quality assurance parameters assure consistent results. The reagents and stains in SelecTech are designed to complement each other to produce a crisp, well delineated hematoxylin and eosin stain.
Catalog No. NC1105685