Learn More
Leica Biosystems HEMATOXYLIN COUNTERSTAIN
Supplier:  Leica Biosystems AR9915
Encompass
Hematoxylin Counterstain is designed to be used in conjunction with BOND Polymer Refine HRP Plex Detection kit (DS9914) and our chromogen reagents. Hematoxylin is the most commonly used nuclear histological counterstain and our Hematoxylin Counterstain provides excellent contrast with a variety of chromogens, especially with our Blue Chromogen (DC9896) and Green Chromogen (DC9913).Hematoxylin Counterstain is supplied in a volume of 15 mL.
Catalog No. NC1940153