Learn More
Selleck Chemical LLC Hematoxylin 5mg 517-28-2 Hydroxybrazilin
SDP
Supplier:  Selleck Chemical LLC S23845MG
Encompass_Preferred
Hematoxylin (Hydroxybrazilin) is a compound that forms strongly colored complexes with certain metal ions, notably Fe(III) and Al(III) salts and a kind of stain in histology. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-544-8