Learn More
Crescent Chemical Co Inc HEMATOXYLIN PURE
SDP
Supplier:  Crescent Chemical Co Inc SE24420.01
Encompass_Preferred
Hematoxylin pure Tested for use in Delafield-Heidenhain staining. Light-sensitive. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 24420.01 CAS 517-28-2
Catalog No. 50-247-707