Learn More
Alkali Scientific Hematoxylin, 100 G
Supplier: Alkali Scientific HEM001CP
Hematoxylin is a histology stain used primarily for tissue staining in medical and research laboratories. It is often combined with eosin to create the Hematoxylin and Eosin (H&E) staining method, which is widely employed for visualizing tissue structures in histological slides. Hematoxylin stains cell nuclei a dark blue or purple, which provides excellent contrast when examining tissue under a microscope. This 100g package of hematoxylin is ideal for laboratories involved in routine histopathology work, offering a high-quality solution for tissue fixation, staining, and examination. It is a crucial reagent for diagnosing diseases, including cancer, by highlighting cellular and tissue abnormalities.