Learn More
Cayman Chemical HemIn chlorIde 1g
SDP
Supplier:  Cayman Chemical 164871 G
Encompass_Preferred
An oxidized form of heme that inhibits eIF2αK1, a repressor of eIF-2α; used experimentally to induce the expression of heme oxygenase-1 in cells and in animals
Catalog No. 50-297-1374