Learn More
Electron Microscopy Sciences Xylene Substitute Hemo-De (d-Limonene) 1 GL
SDP
Supplier:  Electron Microscopy Sciences 2341201
Encompass_Preferred
An intensive study from 1992 throughout 2000 has shown that Hemo-De (d-Limonene) a terpene-based chemical with a pleasant citrus fragrance, was able to replace many of the clinical laboratories toxic reagents, such as carbol-xylene, xylene, ethyl acetate and formalin.
Hemo-De replaces ethyl acetate in the concentration procedure in parasitology; replaces carbol-xylene, xylene and formalin in the trichrome procedure in parasitology.
Hemo-De replaces xylene in most all procedures in histology. Superior solvent and clearing agent that can be used in all tissue processing, deparaffiniating and slide preparation.
Hemo-De is soluble with alcohol and mounting media. It is biodegradable, non-corrosive, non-flammable (Combustible) and contains no benzene and no toluene. Low toxicity levels. There is minimal tissue shrinkage with reasonably fast drying and leaves no residue.
Catalog No. 50-335-75
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.