Learn More
Carbosynth LLC. Purified native Human Hemoglobin A1c protein
SDP
Supplier:  Carbosynth LLC. 30RAH012X1MG
Encompass_Preferred
Glycosylated haemoglobin (HbA1c) levels are used as an indicator of blood glucose in diabetic patients. It is formed by the conjugation of a hexose sugar to the beta-chain of haemoglobin A. HbA1C builds up in red blood cells throughout their lifetime and its levels therefore represent a record of blood glucose levels over the previous 8 weeks or so. The subunit has mol wt of 18 kDa.
Catalog No. 50-125-3199