Learn More
Carbosynth LLC. Hemoglobin Powder (bovine) , 1 Kg
SDP
Supplier:  Carbosynth LLC. 97R1004S100G
Encompass_Preferred
Hemoglobin is a protein that is carried by red cells. It picks up oxygen in the lungs and delivers it to the peripheral tissues to maintain the viability of cells. Hemoglobin is composed of one globin plus four hemes; heme consists of protoporphyrin IX and ferrous iron.
Catalog No. 50-187-0942