Learn More
Medchemexpress LLC Hemopressin (human, mouse) | 1314035-51-2 | 99.95% | 1054.24 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP109110MG
Encompass_Preferred
Hemopressin is a nonapeptide derived from the α1-chain of hemoglobin, originally isolated from rat brain homogenates. It acts as an orally active, selective, and inverse agonist of CB1 cannabinoid receptors, demonstrating antinociceptive action in inflammatory pain models.
  • Derived from the α1-chain of hemoglobin
  • Orally active and selective
  • Inverse agonist of CB1 cannabinoid receptors
  • Exhibits antinociceptive action in inflammatory pain models
Catalog No. 50-004-22746