Learn More
Fine Science Tools Kelly Hemostats, Stainless Steel, Straight, 14 cm
SDP
Supplier:  Fine Science Tools 1301814
Encompass
Tip Shape: Straight/ Tips: Serrated/ Tip Width: 1.5mm/ Clamping Length: 22mm/ Alloy / Material: Stainless Steel/ Length: 14cm
Catalog No. NC9079872