Learn More
Medchemexpress LLC Heneicosane | 629-94-7 | MFCD00009346 | 99.7% | 296.57 | C21H44 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW08984510G
Encompass_Preferred
Heneicosane is a straight-chain saturated hydrocarbon (C21) supplied as an analytical standard for research and analytical applications. It is commonly used as a reference compound in chromatographic and mass spectrometric analyses, and is also reported as a component of insect pheromones.
  • Analytical standard for chromatographic and mass spectrometry methods.
  • High purity, typically 99.7%.
  • Molecular weight 296.57 g/mol and formula C21H44.
  • Available in multiple package sizes, including 10 G.
  • White waxy solid at room temperature; stable and easy to store.
Catalog No. 50-002-08760