Learn More
Cayman Chemical Heneicosapentaenoic Acid
SDP
Supplier:  Cayman Chemical 1067025 MG
Encompass_Preferred
A 21:5 ω-3 fatty acid present in trace amounts in the green alga B. pennata and in fish oils; can be used to study the significance of the position of the double bonds in ω-3 fatty acids; rapidly inactivates PGHS (COX)
Catalog No. 50-289-1052