Learn More
Cayman Chemical HentrIacontn 500mg
SDP
Supplier:  Cayman Chemical 29776500 MG
Encompass_Preferred
An alkane with anti-inflammatory activity; reduces LPS-induced TNF-α, IL-6, and IL-1β production in RAW 264.7 cells; reduces LPS-induced TNF-α, IL-6 and IL-1β production in whole blood and carrageenan-induced paw edema in mice
Catalog No. 50-298-8534