Learn More
Instech Sterile Cather Lock Solution, Heparin Sodium (250 USP units/mL) in Normal Saline (0.9% NaCl) 1 mL in 1mL Syringe, PinPort™ Injector, 50pcs
SDP
Supplier:  Instech USPHS2501NJ1P50
Encompass
Pharmaceutical-grade catheter lock solution prefilled into syringes following the USP <797> standard for compounding sterile preparations. The solution contains heparin sodium (250 USP units/mL) in saline (0.9% NaCl) USP, sterile grade. Each 1 mL NORM-JECT F 1 slip fit luer syringe is filled to 1 mL and capped with a PinPort™ Injector. The syringes are sealed to maintain sterility of the contents and provided in quantities of 50 (10 packs of 5).
Unused prefilled syringes may be stored for up to one (1) year from date of manufacture before expiring. The expiration date is listed on the product label.
For laboratory animal research only. Not approved for human use.
  • Preinstalled PinPort™ Injectors; Ready-to-Use with VABs™ and PinPorts™
  • Pharmaceutical-Grade
  • Sterile Filtered
  • Preservative-Free
Catalog No. NC2856065