Learn More
ATCC HepAD38 (HepG2 Derivative) Human Liver Cell Line, Inducible HBV Expression, Adherent, BSL-2, ~4.5x10^6 Cells/Vial
Supplier:  ATCC CRL3561
Encompass
HepAD38 is a human, liver-derived HepG2 derivative engineered for tetracycline-regulated (inducible) expression of the hepatitis B virus genome. It is used for antiviral compound screening and for production of encapsidated HBV DNA when expression is induced.
  • Inducibly expresses the hepatitis B virus genome under tetracycline control.
  • Suitable for antiviral screening and HBV DNA production.
  • Adherent, epithelial-like cells derived from human liver.
  • Maintain in DMEM/F12 with 10% fetal bovine serum; include tetracycline to repress expression.
  • Typical vial contains approximately 4.5 x 10^6 cells; store in vapor-phase liquid nitrogen.
  • Biosafety level 2 handling is recommended.
  • Culture at 37°C in 95% air, 5% CO2.
Catalog No. NC2955634