Learn More
Medchemexpress LLC Heparan sulfate | 9050-30-0 | 593.47 (monomer) | C12H19NO20S3 (monomer) | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10191650MG
Encompass_Preferred
Heparan sulfate is a naturally occurring linear glycosaminoglycan used as a research reagent to study growth factor signaling, extracellular matrix interactions, and enzymatic degradation. It is supplied as a purified polysaccharide suitable for biochemical assays and cell-based studies.
  • Naturally occurring linear glycosaminoglycan.
  • Used to study growth factor and extracellular matrix interactions.
  • Soluble in water for assay preparation.
  • Monomer formula C12H19NO20S3; monomer MW 593.47.
  • Available in multiple package sizes including 50 mg.
Catalog No. 50-002-48540