Learn More
Sigma Aldrich Fine Chemicals Biosciences Heparin sodium salt from porcine intestinal mucosa | 9041-08-1 | MFCD00081689 | 250 KU
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H3393250KU
Encompass
Heparin sodium salt from porcine intestinal mucosa has been supplemented in the medium prepared for insemination of oocytes with spermatozoa. It has been used to check the effect of heparin on FGF21 (fibroblast growth factor 21) signaling.
Catalog No. NC0827339