Learn More
LIFESPAN TECHNOLOGIES Heparin ELISA Kit, Competitive assay, Sample Types: Buffer, Urine, LMWH Detection Range: 0.003 - 1.0 μg/mL, UFH Detection Range: 0.001 - 0.3 U/mL, 96-well plate format
Supplier:  LIFESPAN TECHNOLOGIES S1800
Encompass
The Lifespan Technologies Heparin ELISA Kit is a highly sensitive tool designed for the direct measurement of low molecular weight heparin (LMWH) and unfractionated heparin (UFH) levels in various biological fluids. This competitive enzyme-linked immunoassay (ELISA) is particularly optimized for low protein content samples such as buffer or urine, ensuring effective quantification for diverse research applications. Utilizing a heparin binding protein conjugated to horseradish peroxidase (HRP), it generates a colorimetric signal inversely proportional to the concentration of heparin, making it invaluable for pharmacokinetic and pharmacodynamic studies.
  • Direct measurement of heparin for accurate quantification
  • Compatible with low protein content biological fluids
  • Sensitive detection of LMWH and UFH
  • Simplified reagent preparation with stabilized liquid conjugate
Catalog No. NC2602866