Learn More
Bioworld HEPARIN SEPAROPOREAGAROSE
Supplier:  Bioworld 201810133
Encompass
HEPARIN SEPAROPORE (AGAROSE) 4B: Heparin Separopore Affinity matrix with immobilized heparin (from porcine intestinal mucosa) by CNBr activation coupling method.; For affinity purification of coagulation factors such as antithrombin III,...
Catalog No. 50-197-692