Learn More
Selleck Chemical LLC Heparin sodium 200mg 9041-08-1
SDP
Supplier:  Selleck Chemical LLC S1346200MG
Encompass_Preferred
Heparin sodium, a sulfated polysaccharide belonging to the family of glycosaminoglycans, has numerous important biological activities associated with its interaction with diverse proteins. Heparin is used as an anticoagulant primarily through its interaction with AT III by enhancing AT-IIImediated inhibition of blood coagulation factors, including thrombin and factor Xa. Heparin prevents the induction of autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1162