Learn More
Medchemexpress LLC Heparin sodium salt | 9041-08-1 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17567100MG
Encompass_Preferred
Heparin sodium salt (Sodium heparin) is an anticoagulant that reversibly binds to antithrombin III (ATIII), significantly accelerating the rate at which ATIII inactivates coagulation enzymes thrombin factor IIa and factor Xa. It also notably inhibits exosome-cell interactions.
  • Reversibly binds to antithrombin III (ATIII)
  • Accelerates ATIII inactivation of thrombin factor IIa and factor Xa
  • Inhibits exosome-cell interactions
Catalog No. 50-003-87499