Learn More
ATCC Hepatitis A virus; Strain: HM175/18f
Supplier:  ATCC VR1402
Encompass
Hepatitis A virus strain HM175/18f is propagated in FRhK-4 cells (ATCC CRL-1688). The virus was isolated in the agarose of a radioimmunofocus assay in FRhK-4 cells and was subsequently passaged in BS-C-1 cells. The strain has applications in enteric and infectious disease research.
Catalog No. 50-238-4052