Learn More
Medchemexpress LLC Hepatitis B Virus Core (128-140) | 160015-13-4 | 99.6% | 1406.63 | 1 MG

Supplier: Medchemexpress LLC HYP17741MG
Hepatitis B Virus Core (128-140) is a peptide derived from the hepatitis B virus core protein. This product is intended for research use only.
- High purity
- Solid, white to off-white appearance
- Soluble in water at ≥ 100 mg/mL
- Specific amino acid sequence
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.