Learn More
Medchemexpress LLC Hepatocyte growth factor activator (recombinant human) | >95.0% | 50 UG

Supplier: Medchemexpress LLC HYP7638050UG
Recombinant human hepatocyte growth factor activator produced in HEK293 cells with a C-terminal His tag. It catalyzes conversion of inactive hepatocyte growth factor (HGF) into the biologically active heterodimer and is supplied lyophilized for use in in vitro biochemical and cell-based assays investigating HGF signaling and proteolytic activation.
- Recombinant human HGFA expressed in HEK293 cells.
- C-terminal His tag for detection and purification.
- Purity greater than 95.0% by reducing SDS-PAGE.
- Molecular weight approximately 95-110 kDa.
- Lyophilized formulation in PBS (pH 7.4) with 5-8% trehalose.
- Recommended reconstitution not below 100 μg/mL in ddH2O.
- Store at -20°C; aliquot and keep at -80°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.