Learn More
XenoTech,LLC Hepatosure™ OptiThaw Kit
SDP
Supplier:  XenoTech,LLC K8500
Encompass_Preferred
Proprietary cell culture media optimized for thawing Hepatosure Pooled cryopreserved hepatocytes, plus Trypan blue solution to obtain accurate cell yield and viability.
Catalog No. NC1636341