Learn More
ATCC HepatoXcell™ Primary Hepatocyte Plating Medium 1x
Supplier:  ATCC PCS450038
Encompass
HepatoXcell™ Plating Media is a complete, ready-to-use, phenol red free media formulated for the purpose of plating fresh or cryopreserved primary hepatocytes. The plating media containing prequalified fetal bovine serum (FBS) has been optimally designed for primary hepatocyte seeding and successful attachment to collagen I coated plates.
Catalog No. NC3456104