Learn More
Carbosynth LLC. Hepcidin-9 (Mouse), 1mg, MW: (M.W. 1068.27), Molecular formula: C50H73N11O13S
SDP
Supplier:  Carbosynth LLC. PLP3858PI1MG
Encompass_Preferred
Synonyms: H-Asp-Thr-Asn-Phe-Pro-Ile-Cys-Ile-Phe-NH2, Salt Form:, Trifluoroacetate, Storage Temperature: -20°C
Catalog No. 50-168-6618