Learn More
Carbosynth LLC. Hepcidin/LEAP-1 (Human) (Bulk), 1mg, Disulfide bonds: Cys7-Cys23, Cys10-Cys13, Cys11-Cys19, and Cys14- Cys22, MW: (M.W. 2789.4), Molecular formula: C113H170N34O31S9, CAS# [342790-21-0]
SDP
Supplier:  Carbosynth LLC. PLP3771PI1MG
Encompass_Preferred
Synonyms: Asp-Thr-His-Phe-Pro-Ile-Cys-Ile-Phe-Cys-Cys-Gly-Cys-Cys-His-Arg-Ser-Lys-Cys-Gly-Met-Cys-Cys-Lys-Thr, Liver-Expressed Antimicrobial Peptide, Hepcidin-25, PLTR (Putative Liver Tumor Regressor), Application: Liver-Specific Antimicrobial Peptide / Iron-Regulatory Hormone, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: A. Krause, et al., FEBS Lett., 480, 147 (2000). (Original; LEAP-1), C.H. Park, et al., J. Biol. Chem., 276, 7806 (2001). (Original; Hepcidin), T. Gan, et al., J. Physiol. Gastrointest. Liver Physiol, 290, G199 (2006).
Catalog No. 50-168-6600