Learn More
Cayman Chemical HepcIdIn25trIfluoROCTAte 100ug
SDP
Supplier:  Cayman Chemical 34768100 UG
Encompass_Preferred
A peptide hormone; binds to ferroportin, inducing its internalization and degradation, which increases intracellular iron levels and decreases dietary iron absorption and plasma iron levels; bactericidal against multidrug-resistant clinical isolates of Gram-negative and Gram-positive bacteria from 6.25-50 UG/ml; levels are increased in the serum of patients with CKD
Catalog No. 50-291-1055