Learn More
Medchemexpress LLC HEPES | 7365-45-9 | 99.9% | 1 G
SDP
Supplier:  Medchemexpress LLC HYD08571G
Encompass_Preferred
HEPES is a nonvolatile zwitterionic chemical buffering agent broadly applied in cell culture. It is effective at pH 6.8 to 8.2 and is also a potent inducer of lysosome biogenesis.
  • Nonvolatile zwitterionic chemical buffering agent
  • Broadly applied in cell culture
  • Effective at pH 6.8 to 8.2
  • Potent inducer of lysosome biogenesis
Catalog No. 50-004-09955