Learn More
Andwin Scientific HEPES ACID FREE 100 gram
SDP
Supplier:  Andwin Scientific 391338
Encompass_Preferred
Hepes is a solution made to perform as a zwitterionic N-substituted aminosulfonic acid buffer. It is ideal for use in the pH 6.8-8.2 range. This product has a pKa of 7.48 at 25C. This product has a Health Hazard NFPA Rating of 2, a Fire Hazard NFPA Rating of 1, and a Reactivity NFPA Rating of 0. It is packaged and sold in a 100 gram glass bottle.
Catalog No. NC9912907