Learn More
Biotang Inc HEPES Buffered Saline 5X, pH 7.0. 125 mM HEPES, 750 mM Sodium Chloride, pH 7.0. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-550
Supplier:  Biotang Inc BTBB456
Encompass_Preferred
HEPES Buffered Saline 5X, pH 7.0. 125 mM HEPES, 750 mM Sodium Chloride, pH 7.0. Prepared in 18.2 megohms water and filtered through 0.22 micron filter. BB-550.
Catalog No. 50-155-805