Learn More
Boston Bioproducts Inc HEPES Buffer (1 M, pH 7.0) - 250ml
Supplier:  Boston Bioproducts Inc BBH70250ML
Encompass_Preferred
The HEPES [4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid)] Buffer is a versatile Good's zwitterionic buffering agent widely used in various applications in biochemistry and molecular biology. It maintains a stable pH in the range of 6.8 to 8.2, making it ideal for various applications at physiological pH, including cell culture, protein purification, isoelectric focusing, enzyme studies, bioprocessing, and live cell imaging.
Catalog No. NC0349834