Learn More
Sigma Aldrich Fine Chemicals Biosciences HEPES sodium salt solution1L
Supplier:  Sigma Aldrich Fine Chemicals Biosciences H36621L
Encompass
HEPES is a general-purpose zwitterionic buffer which does not bind magnesium calcium manganese(II) or copper(II) ions. HEPES has been classified as Good buffer by Dr. Norman Good and his colleagues in 1966. It is zwitterionic at biological pH and is most effective in the pH range of pH 6.8 to 8.2.
Catalog No. 50-290-2512