Learn More
Eurofins Discoverx PathHunter HepG2 STAT3 Pathway Reporter Cell Line, 2 vials. (93-1144C44)
Supplier:  Eurofins Discoverx 931144C44
Encompass
The PathHunter HepG2 STAT3 Pathway Reporter Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting the STAT3 pathway. This cell line enables assessment of STAT3 pathway stimulation based on changes in the expression of a STAT3 response element-driven reporter protein. This product uses EFC technology. The included cell line expresses ProLabel-tagged reporter protein under the control of a STAT3 pathway-inducible transcriptional response element. Pathway activation induces expression of the ProLabel-tagged protein, allowing detection of EFC signal upon addition of EA-containing detection reagent. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/pathways.
Catalog No. 50-200-2238
Only null left
Add to Cart
Request a Quote