Learn More
AG Scientific Inc HEPPS 25 G
Supplier:  AG Scientific Inc H103025GM
Encompass_Preferred
HEPPS is a biological buffer with a useful pH range of 7.3-8.7. pKa=8.0 at 25°C.
Catalog No. 50-226-8308