Learn More
Medchemexpress LLC Heptacosane | 593-49-7 | MFCD00009862 | 98.0% | 380.73 g/mol | C27H56 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW269368250MG
Encompass_Preferred
Heptacosane is a long-chain alkane (C27H56, MW 380.73 g/mol, CAS 593-49-7) supplied as a ≥98.0% analytical standard for research and chromatographic applications. It is intended as a reference compound for method development, calibration, and identification in GC/GC-MS and other analytical techniques. Follow the product SDS for handling and storage.
  • High purity (98.0%) for analytical reliability.
  • Straight-chain C27 hydrocarbon for reference standards.
  • Suitable for GC and GC-MS method development and calibration.
  • Provided in small quantities for laboratory use.
  • Datasheet and SDS available for handling and safety information.
Catalog No. 50-002-81982