Learn More
Gelest Inc Heptadecafluoro-1,1,2,2-tetrahydrodecyltrichlorosilane, 78560-44-8, MFCD00042344, 5G
Supplier:  Gelest Inc SIH5841.05G
Encompass
This item has a minimum qty of 8 per supplier requirements.
Molecular Formual: C10H4CL3F17SI, Molecular Weight: 581.56, Boiling Point: 216-8, Melting Point: 10-11
Catalog No. NC9312070