Learn More
Medchemexpress LLC Heptadecanoic acid | 506-12-7 | MFCD00002751 | 270.45 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW004284R
Encompass_Preferred
Heptadecanoic acid is an odd-chain saturated fatty acid (OCS-FA) with oral activity. It can inhibit cell proliferation and induce apoptosis. It has antitumor activity and is associated with a number of diseases, including coronary heart disease, pre-diabetes and type 2 diabetes, and multiple sclerosis.
  • Inhibits proliferation and migration of PC-9 and PC-9/GR cells
  • Promotes apoptosis by activating the PI3K/Akt signaling pathway
  • Increases the frequency of defecation in rats
  • Studied for its potential role in lipid metabolism and as a dietary supplement
Catalog No. 50-004-47760