Learn More
Medchemexpress LLC Heptadecanoic acid-d3 | 202528-95-8 | 273.47 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW004284S1MG
Encompass_Preferred
Heptadecanoic acid-d3 is the deuterium labeled Heptadecanoic acid, an odd chain saturated fatty acid (OCS-FA) associated with various diseases such as coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis. This compound is used in research for its unique characteristics:
  • It can be employed as a tracer in research applications.
  • It serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes, including hydrogen and carbon, are incorporated into drug molecules primarily for quantitation during the drug development process.
  • Deuteration of drug molecules has been noted for its potential to influence their pharmacokinetic and metabolic profiles.
Catalog No. 50-004-47986