Learn More
Medchemexpress LLC Heptadecanoic acid-d33 | 352431-41-5 | 303.65 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW004284S150MG
Encompass_Preferred
Heptadecanoic acid-d33 is a deuterium labeled odd chain saturated fatty acid (OCS-FA). It is associated with several diseases, including coronary heart disease, prediabetes, type 2 diabetes, and multiple sclerosis. This product is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules as tracers
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-49176