Learn More
Cayman Chemical HeptadecnoIc AcId 1g
SDP
Supplier:  Cayman Chemical 197221 G
Encompass_Preferred
An odd-chain saturated fatty acid; has been found in milk fat; has been used as an internal standard for the quantification of fatty acids in human plasma by LC- and GC-MS; has been used as a biomarker for dairy fat intake
Catalog No. 50-298-2230