Learn More
Cayman Chemical HeptadecnoIc AcId methyl10g
SDP
Supplier:  Cayman Chemical 2672310 G
Encompass_Preferred
An esterified form of heptadecanoic acid; has been found in biodiesel; has been used as an internal standard for the quantification of fatty acid methyl esters in human plasma
Catalog No. 50-293-4956