Learn More
Cayman Chemical Heptadecnoyl EthnolamIde 10mg
SDP
Supplier:  Cayman Chemical 9034210 MG
Encompass_Preferred
A synthetic analog of PEA which incorporates an odd-numbered (17-carbon) fatty acid achain; potentiates the Ca2+ influx in response to AEA several fold in cells expressing human recombinant VR1
Catalog No. 50-289-6078